Bis(trimethylsilyl)acetylene 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11960-30
A11960-09
A11960-18
CAAAAA11960-30EA
1349.32
CAD
CAAAAA11960-30
CAAAA11960-09
CAAAA11960-18
Bis(trimethylsilyl)acetylene 99%
Bis(trimethylsilyl)acetylene
Fieser: 5,44
Purity: 99%
CAS No.: 14630-40-1
Molecular Formula: C8H18Si2
Formula Weight: 170.40
Flash Point: 2°(35°F)
Refractive Index: 1.4270
Melting Point: 20-24°
Boiling Point: 134-136°
Beilstein Reference: 906870
UN No.: 1993
MDL No.: MFCD00008276
Formula:
C₈H₁₈Si₂ MW: 170.4 g/mol Boiling Pt: 134…136 °C Melting Pt: 20…24 °C Density: 0.770 Flash Pt: 2 °C (35 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008276 CAS Number: 14630-40-1 EINECS: 238-671-9 UN: 1993 ADR: 3 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...