2-Chloroethyl toluene-4-sulfonate 97%

Supplier: Thermo Scientific Chemicals
Warning

A11863-14 A11863-22 A11863-36
CAAAA11863-14EA 90.38 CAD
CAAAA11863-14 CAAAA11863-22 CAAAA11863-36
2-Chloroethyl toluene-4-sulfonate 97%
2-Chloroethyl toluene-4-sulphonate

Purity: 97% CAS No.: 80-41-1 Molecular Formula: C9H11ClO3S Formula Weight: 234.70 Storage Temperature: 2°C to 8°C Flash Point: 110°(230°F) Refractive Index: 1.5290 Boiling Point: 158-159°/0.5mm Beilstein Reference: 1968816 MDL No.: MFCD00000970

Formula: C₉H₁₁ClO₃S
Boiling Pt: 158…159 °C/0.5 mm
Density: 1.294
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00000970
CAS Number: 80-41-1
EINECS: 201-277-2
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR