Acetophenone oxime 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Methyl phenyl ketoxime, 1-Phenyl-1-ethanone oxime, Acetophenone Oxime

A11804-09 A11804-18 A11804-30
CAAAA11804-09EA 69.99 CAD
CAAAA11804-09 CAAAA11804-18 CAAAA11804-30
Acetophenone oxime 98%
Acetophenone oxime

Used as a ligand in transition-metal complex catalyst chemistry. Oxime acts as an antioxidant, radical scavenger which find applications in textile, plastic, paint, detergent, and rubber industry.


Purity: 98% CAS No.: 613-91-2 Molecular Formula: C8H9NO Formula Weight: 135.16 Storage Temperature: -30°C to -10°C Melting Point: 58-60° Boiling Point: 118-120°/20mm Beilstein Reference: 1562059 MDL No.: MFCD00013931

Formula: C₈H₉NO
MW: 135.17 g/mol
Boiling Pt: 118…120 °C/20 mm
Melting Pt: 58…60 °C
Storage Temperature: Freezer
MDL Number: MFCD00013931
CAS Number: 613-91-2
EINECS: 210-360-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR