(4-Chlorobenzyl)triphenylphosphonium chloride 98+%

Supplier: Thermo Scientific Chemicals
Warning

A11769-36 A11769-14 A11769-22
CAAAAA11769-36EA 1004.88 CAD
CAAAAA11769-36 CAAAA11769-14 CAAAA11769-22
(4-Chlorobenzyl)triphenylphosphonium chloride 98+%
(4-Chlorobenzyl)triphenylphosphonium chloride

Purity: 98+% CAS No.: 1530-39-8 Molecular Formula: C25H21Cl2P Formula Weight: 423.32 Melting Point: 286-291° Beilstein Reference: 4173400

Formula: C₂₅H₂₁Cl₂P
Melting Pt: 286…291 °C
Storage Temperature: Ambient
MDL Number: MFCD00041533
CAS Number: 1530-39-8
EINECS: 216-228-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR