Fluorescein disodium salt hydrate

Supplier: Thermo Scientific Chemicals

A11659-0E A11659-22 A11659-36
CAAAA11659-0EEA 486.64 CAD
CAAAA11659-0E CAAAA11659-22 CAAAA11659-36
Fluorescein disodium salt hydrate
Fluorescein disodium salt hydrate

Fluorescein disodium salt hydrate is used in dye, ink pigment, in cosmetic products, to detect corneal epithelial defects, and also used as a fluorescent tracer to study blood-brain barrier.


CAS No.: 518-47-8 Molecular Formula: C20H10Na2O5.xH2O Formula Weight: 376.28 Odor: Odorless pH: 8.3 Melting Point: 250° Beilstein Reference: 3833041 Merck Reference: 14,4159 MDL No.: MFCD00167039

Formula: C₂₀H₁₀Na₂O₅
Melting Pt: >250 °C
Density: 1.601 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00167039
CAS Number: 518-47-8
EINECS: 208-253-0
Merck Index: 14,4159
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=949f2473a2f0c6f8d00fd80ca2f80b730bc8076a&vid=fe174603ab8c1727364c1e09d665a1ad5a6d9106&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR