3-Amino-5-methyl-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Amino-5-methylpyrazole, 5-Methyl-1H-pyrazol-3-amine

A11642-06 A11642-14 A11642-22
CAAAAA11642-06EA 54.04 CAD
CAAAAA11642-06 CAAAAA11642-14 CAAAAA11642-22
3-Amino-5-methyl-1H-pyrazole 97%
3-Amino-5-methyl-1H-pyrazole

Purity: 97% CAS No.: 31230-17-8 Molecular Formula: C4H7N3 Formula Weight: 97.12 Flash Point: 110°(230°F) Melting Point: 42-47° Boiling Point: 212-213°/14mm Beilstein Reference: 1904 MDL No.: MFCD00075180

Formula: C₄H₇N₃
MW: 97.12 g/mol
Boiling Pt: 212…213 °C/14 mm
Melting Pt: 42…47 °C
Flash Pt: >110 °C (230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00075180
CAS Number: 31230-17-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR