4,4'-Dimethoxytrityl chloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A11626-06 A11626-14 A11626-22
CAAAA11626-06EA 79.95 CAD
CAAAA11626-06 CAAAA11626-14 CAAAA11626-22
4,4'-Dimethoxytrityl chloride 98%
4,4'-Dimethoxytrityl chloride

Purity: 98% CAS No.: 40615-36-9 Molecular Formula: C21H19ClO2 Formula Weight: 338.83 Melting Point: 121-127° Beilstein Reference: 2471942

Formula: C₂₁H₁₉ClO₂
Melting Pt: 121…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00008409
CAS Number: 40615-36-9
EINECS: 255-002-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR