Diphenylphosphinic acid 99%
Supplier: Thermo Scientific Chemicals
A11563-06
A11563-14
A11563-22
CAAAA11563-06EA
71.73
CAD
CAAAA11563-06
CAAAA11563-14
CAAAA11563-22
Diphenylphosphinic acid 99%
Diphenylphosphinic acid
Purity: 99%
CAS No.: 1707-03-5
Molecular Formula: C12H11O2P
Formula Weight: 218.19
Melting Point: 193-197°
Beilstein Reference: 2804567
Formula:
C₁₂H₁₁O₂P Melting Pt: 193…197 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002132 CAS Number: 1707-03-5 EINECS: 216-948-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...