Triphenylphosphine dibromide 96%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11552-09
A11552-18
A11552-30
CAAAA11552-09EA
85.48
CAD
CAAAA11552-09
CAAAA11552-18
CAAAA11552-30
Triphenylphosphine dibromide 96%
Triphenylphosphine dibromide
Fieser: 1,1247 13,333
Purity: 96%
CAS No.: 1034-39-5
Molecular Formula: (C6H5)3PBr2
Formula Weight: 422.10
Melting Point: ca 260° dec.
Beilstein Reference: 917652
UN No.: 3261
MDL No.: MFCD00000054
Formula:
C₁₈H₁₅Br₂P Melting Pt: ca 260 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00000054 CAS Number: 1034-39-5 EINECS: 213-855-1 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...