3-Amino-2-cyclohexen-1-one, (max. 5% H₂O) 98% (dry weight)
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11474-06
A11474-14
CAAAA11474-03EA
25.96
CAD
CAAAA11474-03
CAAAA11474-06
CAAAA11474-14
3-Amino-2-cyclohexen-1-one, (max. 5% H₂O) 98% (dry weight)
3-Amino-2-cyclohexen-1-one
Purity: 98%
CAS No.: 5220-49-5
Molecular Formula: C6H9NO
Formula Weight: 111.15
Melting Point: 128-133°(dried)
Beilstein Reference: 471354
Formula:
C₆H₉NO Boiling Pt: 211 °C (1013 hPa) Melting Pt: 128…133 °C (dried) Storage Temperature: Ambient |
MDL Number:
MFCD00013783 CAS Number: 5220-49-5 EINECS: 226-014-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...