2-Furoic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Furan-2-carboxylic acid, 2-Furancarboxylic acid
A11454-22
A11454-36
CAAAA11454-22EA
60.04
CAD
CAAAA11454-22
CAAAA11454-36
2-Furoic acid 98%
2-Furoic acid
Purity: 98%
CAS No.: 88-14-2
Molecular Formula: C5H4O3
Formula Weight: 112.08
Flash Point: 137°(278°F)
Odor: Odorless
Boiling Point: 230-232° subl.
Beilstein Reference: 110149
Merck Reference: 14,4307
MDL No.: MFCD00003238
Formula:
C₅H₄O₃ MW: 112.08 g/mol Boiling Pt: 230…232 °C (subl.) Melting Pt: 127…132 °C Flash Pt: 137 °C(278 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00003238 CAS Number: 88-14-2 EINECS: 201-803-0 Merck Index: 14,4307 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...