2,5-Dihydroxy-p-benzoquinone 98%

Supplier: Thermo Scientific Chemicals
Warning

A11442-06 A11442-14
CAAAAA11442-06EA 42.98 CAD
CAAAAA11442-06 CAAAAA11442-14
2,5-Dihydroxy-p-benzoquinone 98%
2,5-Dihydroxy-p-benzoquinone

Purity: 98% CAS No.: 615-94-1 Molecular Formula: C6H4O4 Formula Weight: 140.10 Melting Point: ca 220° subl. Beilstein Reference: 1939229

Formula: C₆H₄O₄
Melting Pt: ca 220 °C (subl.)
Storage Temperature: Ambient
MDL Number: MFCD00001598
CAS Number: 615-94-1
EINECS: 210-454-3
UN: 2811
ADR: "6.1",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR