1,3-Phenylenediacetic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11429-04
A11429-09
A11429-14
CAAAAA11429-04EA
54.83
CAD
CAAAAA11429-04
CAAAAA11429-09
CAAAA11429-14
1,3-Phenylenediacetic acid 97%
1,3-Phenylenediacetic acid
Purity: 97%
CAS No.: 19806-17-8
Molecular Formula: C10H10O4
Formula Weight: 194.19
Odor: Odorless
Melting Point: 172-178°
Beilstein Reference: 2616749
MDL No.: MFCD00004341
Formula:
C₁₀H₁₀O₄ Melting Pt: 172…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00004341 CAS Number: 19806-17-8 EINECS: 243-332-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...