Pyridine-2-carbaldehyde 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11391-14
A11391-22
A11391-36
CAAAAA11391-14EA
62.25
CAD
CAAAAA11391-14
CAAAA11391-22
CAAAA11391-36
Pyridine-2-carbaldehyde 99%
Pyridine-2-carbaldehyde
Purity: 99%
CAS No.: 1121-60-4
Molecular Formula: C6H5NO
Formula Weight: 107.11
Storage Temperature: 2°C to 8°C
Flash Point: 68°(154°F)
Refractive Index: 1.5360
Odor: Characteristic
pH: 6.5
Melting Point: -21°
Boiling Point: 180-181°
Beilstein Reference: 105341
UN No.: 2810
MDL No.: MFCD00006290
Formula:
C₆H₅NO Boiling Pt: 180…181 °C Melting Pt: –21 °C Density: 1.124 Flash Pt: 68 °C(154 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006290 CAS Number: 1121-60-4 EINECS: 214-333-6 UN: 2810 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...