Naphthalene black 10B (naphthol blue black, acid black 1)
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Sodium 4-amino-5-hydroxy-3-((4-nitrophenyl)diazenyl)-6-(phenyldiazenyl)naphthalene-2,7-disulphonate, Amido Black 10B, Sodium 4-amino-5-hydroxy-3-((4-nitrophenyl)diazenyl)-6-(phenyldiazenyl)naphthalene-2,7-disulfonate
A11374-14
A11374-22
CAAAA11374-14EA
92.6
CAD
CAAAA11374-14
CAAAA11374-22
Naphthalene black 10B (naphthol blue black, acid black 1)
Naphthalene Black 10B (Naphthol Blue Black, Acid Black 1)
CAS No.: 1064-48-8
Molecular Formula: C22H14N6Na2O9S2
Formula Weight: 616.49
Odor: Odorless
pH: 9.0
Beilstein Reference: 5374263
Formula:
C₂₂H₁₄N₆Na₂O₉S₂ MW: 616.49 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00004017 CAS Number: 1064-48-8 EINECS: 213-903-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...