o-Xylene 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1,2-Dimethylbenzene

A11358-AP A11358-AE A11358-AE A11358-0F A11358-0F A11358-AP
CAAAAA11358-0FEA 191.18 CAD
AAA11358-AE CAAAAA11358-0F AAAA11358-0F CAAAA11358-AE CAAAAA11358-AP AAAA11358-AP
o-Xylene 99%
o-Xylene
Formula: C₈H₁₀
MW: 106.17 g/mol
Boiling Pt: 143…145 °C
Melting Pt: –25 °C
Density: 0.877
Flash Pt: 32 °C (89 °F)
MDL Number: MFCD00008519
CAS Number: 95-47-6
UN: 1307
ADR: 3,III
Merck Index: 14,10081
https://ca.vwr.com/stibo/web/std.lang.all/85/44/768544.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR