2,6-Dimethyl-p-benzoquinone 99%

Supplier: Thermo Scientific Chemicals
Warning

A11342-03 A11342-06 A11342-14
CAAAA11342-03EA 66.52 CAD
CAAAA11342-03 CAAAA11342-06 CAAAA11342-14
2,6-Dimethyl-p-benzoquinone 99%
2,6-Dimethyl-p-benzoquinone

Purity: 99% CAS No.: 527-61-7 Molecular Formula: C8H8O2 Formula Weight: 136.15 Melting Point: 69-71° Beilstein Reference: 2041345

Formula: C₈H₈O₂
Melting Pt: 69…71 °C
Storage Temperature: Ambient
MDL Number: MFCD00001605
CAS Number: 527-61-7
EINECS: 208-420-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR