Salicylaldoxime 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11080-14
A11080-22
A11080-36
CAAAA11080-14EA
58.62
CAD
CAAAA11080-14
CAAAA11080-22
CAAAA11080-36
Salicylaldoxime 98%
Salicylaldoxime
Purity: 98%
CAS No.: 94-67-7
Molecular Formula: C7H7NO2
Formula Weight: 137.14
Melting Point: 55-59°
Beilstein Reference: 1859881
Merck Reference: 14,8327
MDL No.: MFCD00002120
Formula:
C₇H₇NO₂ Melting Pt: 55…59 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002120 CAS Number: 94-67-7 EINECS: 202-353-8 Merck Index: 14,8327 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...