Tetrabutylammonium trifluoromethanesulfonate 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11013-06
A11013-14
A11013-22
CAAAA11013-06EA
91.32
CAD
CAAAA11013-06
CAAAA11013-14
CAAAA11013-22
Tetrabutylammonium trifluoromethanesulfonate 99%
Tetrabutylammonium trifluoromethanesulphonate
Purity: 99%
CAS No.: 35895-70-6
Molecular Formula: [CH3(CH2)3]4NCF3SO3
Formula Weight: 391.54
Melting Point: 112-115°
Beilstein Reference: 4172978
Formula:
C₁₇H₃₆F₃NO₃S Melting Pt: 112…115 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042585 CAS Number: 35895-70-6 EINECS: 252-783-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...