3-Amino-4-bromo-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

A11012-06 A11012-14
CAAAA11012-03EA 100.55 CAD
CAAAA11012-03 CAAAA11012-06 CAAAA11012-14
3-Amino-4-bromo-1H-pyrazole 97%
3-Amino-4-bromo-1H-pyrazole

Purity: 97% CAS No.: 16461-94-2 Molecular Formula: C3H4BrN3 Formula Weight: 161.99 Odor: Odorless Melting Point: 134-137° Beilstein Reference: 1617109 MDL No.: MFCD00082728

Formula: C₃H₄BrN₃
Melting Pt: 134…137 °C
Storage Temperature: Ambient
MDL Number: MFCD00082728
CAS Number: 16461-94-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR