Methyl-3,5-dibromo-4-hydroxybenzoate 98%
Supplier: Thermo Scientific Chemicals
A10799-09
A10799-18
A10799-30
CAAAA10799-09EA
46.14
CAD
CAAAA10799-09
CAAAA10799-18
CAAAA10799-30
Methyl-3,5-dibromo-4-hydroxybenzoate 98%
Methyl 3,5-dibromo-4-hydroxybenzoate
Purity: 98%
CAS No.: 41727-47-3
Molecular Formula: C8H6Br2O3
Formula Weight: 309.94
Melting Point: 123-125°
Beilstein Reference: 2373854
Formula:
C₈H₆Br₂O₃ Melting Pt: 123…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016416 CAS Number: 41727-47-3 EINECS: 255-521-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...