Ethyl-2-amino-4-methylthiophene-3-carboxylate 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-amino-4-methylthiophene-3-carboxylate

A10790-06 A10790-14
CAAAA10790-03EA 125.18 CAD
CAAAA10790-03 CAAAA10790-06 CAAAA10790-14
Ethyl-2-amino-4-methylthiophene-3-carboxylate 98+%
Ethyl 2-amino-4-methylthiophene-3-carboxylate

Purity: 98+% CAS No.: 43088-42-2 Molecular Formula: C8H11NO2S Formula Weight: 185.25 Melting Point: 79-83° Beilstein Reference: 1366073

Formula: C₈H₁₁NO₂S
MW: 185.25 g/mol
Melting Pt: 79…83 °C
Storage Temperature: Ambient
MDL Number: MFCD00051669
CAS Number: 43088-42-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR