Isonicotinaldehyde 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10656-14
A10656-22
A10656-36
CAAAA10656-14EA
74.42
CAD
CAAAA10656-14
CAAAA10656-22
CAAAA10656-36
Isonicotinaldehyde 97%
Isonicotinaldehyde
Fieser: 11,448
Purity: 97%
CAS No.: 872-85-5
Molecular Formula: C6H5NO
Formula Weight: 107.11
Storage Temperature: 2°C to 8°C
Flash Point: 75°(167°F)
Refractive Index: 1.5440
Melting Point: -4°
Boiling Point: 198°
Beilstein Reference: 105342
MDL No.: MFCD00006425
Formula:
C₆H₅NO Boiling Pt: 198 °C Melting Pt: –4 °C Density: 1.132 Flash Pt: 75 °C(167 °F) Storage Temperature: Refrigerator |
MDL Number:
MFCD00006425 CAS Number: 872-85-5 EINECS: 212-832-3 UN: 1760 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...