α-Benzoinoxime 98+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Hydroxy-1,2-diphenylethanone oxime, α-Benzoin oxime

A10630-14 A10630-22 A10630-36
CAAAA10630-14EA 68.26 CAD
CAAAA10630-14 CAAAA10630-22 CAAAA10630-36
α-Benzoinoxime 98+%
α-Benzoinoxime

Purity: 98+% CAS No.: 441-38-3 Molecular Formula: C14H13NO2 Formula Weight: 227.26 Melting Point: 151-154° Beilstein Reference: 2051941 Merck Reference: 14,1094 MDL No.: MFCD00004501

Formula: C₁₄H₁₃NO₂
MW: 227.26 g/mol
Melting Pt: 151…154 °C
Storage Temperature: Ambient
MDL Number: MFCD00004501
CAS Number: 441-38-3
EINECS: 207-127-2
Merck Index: 14,1094

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR