5-Nitro-2-furoic acid 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Nitrofuran-2-carboxylic acid
A10597-22
A10597-06
A10597-14
CAAAAA10597-22EA
690.46
CAD
CAAAAA10597-22
CAAAA10597-06
CAAAA10597-14
5-Nitro-2-furoic acid 98+%
5-Nitro-2-furoic acid
Purity: 98+%
CAS No.: 645-12-5
Molecular Formula: C5H3NO5
Formula Weight: 157.08
Melting Point: 186-188°
Beilstein Reference: 139373
Formula:
C₅H₃NO₅ Melting Pt: 186…188 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003240 CAS Number: 645-12-5 EINECS: 211-432-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...