Dimethyl-2-aminoterephthalate 99%

Supplier: Thermo Scientific Chemicals
Warning

A10567-0B A10567-18 A10567-30
CAAAAA10567-0BEA 906.92 CAD
CAAAAA10567-0B CAAAA10567-18 CAAAA10567-30
Dimethyl-2-aminoterephthalate 99%
Dimethyl 2-aminoterephthalate

Purity: 99% CAS No.: 5372-81-6 Molecular Formula: C10H11NO4 Formula Weight: 209.20 Melting Point: 130-134° Beilstein Reference: 2808505

Formula: C₁₀H₁₁NO₄
Melting Pt: 130…134 °C
Storage Temperature: Ambient
MDL Number: MFCD00008427
CAS Number: 5372-81-6
EINECS: 226-364-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR