4-Tritylaniline 97%
Supplier: Thermo Scientific Chemicals
Synonyms:
4-(Triphenylmethyl)aniline
A10550-06
A10550-14
CAAAA10550-06EA
68.26
CAD
CAAAA10550-06
CAAAA10550-14
4-Tritylaniline 97%
4-Tritylaniline
Purity: 97%
CAS No.: 22948-06-7
Molecular Formula: C25H21N
Formula Weight: 335.45
Melting Point: 256-260°
Beilstein Reference: 2750328
Formula:
C₂₅H₂₁N Melting Pt: 256…260 °C Storage Temperature: Ambient |
MDL Number:
MFCD00007898 CAS Number: 22948-06-7 EINECS: 245-346-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...