4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10545-09
A10545-18
A10545-30
CAAAA10545-09EA
74.58
CAD
CAAAA10545-09
CAAAA10545-18
CAAAA10545-30
4-Methoxytrityl chloride (p-anisylchlorodiphenylmethane) 97%
4-Methoxytrityl chloride (p-Anisylchlorodiphenylmethane)
Purity: 97%
CAS No.: 14470-28-1
Molecular Formula: C20H17ClO
Formula Weight: 308.81
Melting Point: 121-125°
Beilstein Reference: 798425
Formula:
C₂₀H₁₇ClO Melting Pt: 121…125 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000814 CAS Number: 14470-28-1 EINECS: 238-463-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...