Methyl-3-methyl-2-furoate 98%
Supplier: Thermo Scientific Chemicals
A10541-09
A10541-06
A10541-14
CAAAAA10541-09EA
317.58
CAD
CAAAAA10541-09
CAAAA10541-06
CAAAA10541-14
CAAAA10541-22
Methyl-3-methyl-2-furoate 98%
Methyl 3-methyl-2-furoate
Purity: 98%
CAS No.: 6141-57-7
Molecular Formula: C7H8O3
Formula Weight: 140.14
Flash Point: 82°(180°F)
Melting Point: 33-37°
Boiling Point: 60°/1mm
Beilstein Reference: 116835
MDL No.: MFCD00014110
Formula:
C₇H₈O₃ Boiling Pt: 60 °C/1 mm Melting Pt: 33…37 °C Flash Pt: 82 °C(180 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00014110 CAS Number: 6141-57-7 EINECS: 228-131-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...