4,6-Dihydroxy-2-methylpyrimidine 99%

Supplier: Thermo Scientific Chemicals

A10535-09 A10535-18
CAAAA10535-09EA 95.27 CAD
CAAAA10535-09 CAAAA10535-18
4,6-Dihydroxy-2-methylpyrimidine 99%
4,6-Dihydroxy-2-methylpyrimidine

Purity: 99% CAS No.: 40497-30-1 Molecular Formula: C5H6N2O2 Formula Weight: 126.12 Melting Point: 360° Beilstein Reference: 115646

Formula: C₅H₆N₂O₂
Melting Pt: >360 °C
Storage Temperature: Ambient
MDL Number: MFCD00006104
CAS Number: 40497-30-1
EINECS: 254-941-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR