Chloranilic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10493-14 A10493-22 A10493-36
CAAAA10493-14EA 69.68 CAD
CAAAA10493-14 CAAAA10493-22 CAAAA10493-36
Chloranilic acid 98+%
Chloranilic acid

Purity: 98+% CAS No.: 87-88-7 Molecular Formula: C6H2Cl2O4 Formula Weight: 208.99 Melting Point: 300° dec. Beilstein Reference: 1875040 Merck Reference: 14,2079 MDL No.: MFCD00001596

Formula: C₆H₂Cl₂O₄
Melting Pt: >300 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00001596
CAS Number: 87-88-7
EINECS: 201-780-7
Merck Index: 14,2079

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR