3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10482-03
A10482-06
CAAAA10482-03EA
61.94
CAD
CAAAA10482-03
CAAAA10482-06
3-Hydroxy-4-methoxycinnamic acid predominantly trans ≥98%
3-Hydroxy-4-methoxycinnamic acid
Purity: 98+%
CAS No.: 537-73-5
Molecular Formula: C10H10O4
Formula Weight: 194.19
Melting Point: ca 231° dec.
Beilstein Reference: 2212760
Formula:
C₁₀H₁₀O₄ Melting Pt: ∼ 231 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00004391 CAS Number: 537-73-5 EINECS: 208-676-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...