Methyl-6-chloronicotinate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10396-06
A10396-14
CAAAA10396-03EA
59.93
CAD
CAAAA10396-03
CAAAA10396-06
CAAAA10396-14
Methyl-6-chloronicotinate 98%
Methyl 6-chloronicotinate
Purity: 98%
CAS No.: 73781-91-6
Molecular Formula: C7H6ClNO2
Formula Weight: 171.58
Melting Point: 83-88°
Beilstein Reference: 128648
Formula:
C₇H₆ClNO₂ Melting Pt: 83…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023420 CAS Number: 73781-91-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...