4-Methylumbelliferone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A10337-0B
A10337-18
A10337-30
CAAAA10337-0BEA
576.7
CAD
CAAAA10337-0B
CAAAA10337-18
CAAAA10337-30
4-Methylumbelliferone 97%
4-Methylumbelliferone
Soluble in alcohol. Insoluble in cold water
Purity: 97%
CAS No.: 90-33-5
Molecular Formula: C10H8O3
Formula Weight: 176.17
Melting Point: 186-191°
Solubility: Soluble in alcohol. Insoluble in cold water
Beilstein Reference: 142217
Merck Reference: 14,4854
MDL No.: MFCD00006866
Soluble in alcohol. Insoluble in cold water
Formula:
C₁₀H₈O₃ Melting Pt: 186…191 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006866 CAS Number: 90-33-5 EINECS: 201-986-7 Merck Index: 14,4854 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...