Sodium 3-nitrobenzenesulfonate, (max. 2% H₂O) 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Sodium 3-nitrobenzenesulphonate, 3-Nitrobenzenesulphonic acid, sodium salt, 3-Nitrobenzenesulfonic acid, sodium salt

A10244-0E A10244-22 A10244-36
CAAAA10244-0EEA 524.56 CAD
CAAAA10244-0E CAAAA10244-22 CAAAA10244-36
Sodium 3-nitrobenzenesulfonate, (max. 2% H₂O) 98%
Sodium 3-nitrobenzenesulphonate

Purity: 98% CAS No.: 127-68-4 Molecular Formula: C6H4NNaO5S.xH2O Formula Weight: 225.16(anhy) Melting Point: 350° MDL No.: MFCD00007490

Formula: C₆H₄NNaO₅S
Melting Pt: 350 °C
Storage Temperature: Ambient
MDL Number: MFCD00007490
CAS Number: 127-68-4
EINECS: 204-857-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR