2-Picolyl chloride hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10226-06
A10226-14
A10226-22
CAAAA10226-06EA
63.2
CAD
CAAAA10226-06
CAAAA10226-14
CAAAA10226-22
2-Picolyl chloride hydrochloride 98%
2-Picolyl chloride hydrochloride
Purity: 98%
CAS No.: 6959-47-3
Molecular Formula: C6H6ClN.HCl
Formula Weight: 164.04
Melting Point: 120-124°
Beilstein Reference: 3689155
UN No.: 3261
MDL No.: MFCD00012811
Formula:
C₆H₇Cl₂N Melting Pt: 120…124 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012811 CAS Number: 6959-47-3 EINECS: 230-149-9 UN: 3261 ADR: 8,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...