Diethyl (cyanomethyl)phosphonate 98+%

Supplier: Thermo Scientific Chemicals
Warning

A10218-14 A10218-22 A10218-36
CAAAA10218-14EA 176.96 CAD
CAAAA10218-14 CAAAA10218-22 CAAAA10218-36
Diethyl (cyanomethyl)phosphonate 98+%
Diethyl (cyanomethyl)phosphonate

Fieser: 1,250 2,130 3,96 4,150


Purity: 98+% CAS No.: 2537-48-6 Molecular Formula: C6H12NO3P Formula Weight: 177.14 Flash Point: 110°(230°F) Refractive Index: 1.4330 Boiling Point: 132-135°/1.2mm Beilstein Reference: 1772981 UN No.: 3278 MDL No.: MFCD00001893

Formula: C₆H₁₂NO₃P
Boiling Pt: 132…135 °C/1.2 mm
Density: 1.094
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00001893
CAS Number: 2537-48-6
EINECS: 219-806-0
UN: 3278
ADR: "6.1",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR