Phthalan 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,3-Dihydrobenzo[c]furan

A10217-06 A10217-14 A10217-22
CAAAA10217-06EA 89.59 CAD
CAAAA10217-06 CAAAA10217-14 CAAAA10217-22
Phthalan 98%
Phthalan

Purity: 98% CAS No.: 496-14-0 Molecular Formula: C8H8O Formula Weight: 120.15 Flash Point: 63°(145°F) Refractive Index: 1.5460 Boiling Point: 192° Beilstein Reference: 111932 MDL No.: MFCD00005932

Formula: C₈H₈O
MW: 120.15
Boiling Pt: 192 °C
Density: 1.094
Flash Pt: 63 °C(145 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005932
CAS Number: 496-14-0
EINECS: 207-815-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR