Benzoin 99%

Supplier: Thermo Scientific Chemicals

Synonyms: 2-Hydroxy-2-phenylacetophenone

A10188-0B A10188-0I A10188-30
CAAAA10188-0BEA 164.32 CAD
CAAAA10188-0B CAAAA10188-0I CAAAA10188-30
Benzoin 99%
Benzoin

Fieser: 6,34


Purity: 99% CAS No.: 119-53-9 Molecular Formula: C14H12O2 Formula Weight: 212.25 Flash Point: 181°(357°F) Odor: Pleasant Boiling Point: 334-344° Beilstein Reference: 391839 Merck Reference: 14,1093 MDL No.: MFCD00004496

Formula: C₁₄H₁₂O₂
MW: 212.25 g/mol
Boiling Pt: 334…344 °C
Melting Pt: 131…136 °C
Density: 1.310
Flash Pt: 181 °C(357 °F)
Storage Temperature: Ambient
MDL Number: MFCD00004496
CAS Number: 119-53-9
EINECS: 204-331-3
Merck Index: 14,1093

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR