1-(2-Aminoethyl)piperazine 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A10154-0B
A10154-30
CAAAA10154-0BEA
167.48
CAD
CAAAA10154-0B
CAAAA10154-30
1-(2-Aminoethyl)piperazine 98%
1-(2-Aminoethyl)piperazine
Purity: 98%
CAS No.: 140-31-8
Molecular Formula: C6H15N3
Formula Weight: 129.21
Flash Point: 93°(199°F)
Refractive Index: 1.5000
Melting Point: -19°
Boiling Point: 222°
Beilstein Reference: 104363
UN No.: 2815
MDL No.: MFCD00005971
Formula:
C₆H₁₅N₃ Boiling Pt: 222 °C Melting Pt: –19 °C Density: 0.983 Flash Pt: 93 °C(199 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005971 CAS Number: 140-31-8 EINECS: 205-411-0 UN: 2815 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...