Aluminium tri-tert-butanolate ≥97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
89891-06
89891-14
89891-22
CAAA89891-06EA
76.16
CAD
CAAA89891-06
CAAA89891-14
CAAA89891-22
Aluminium tri-tert-butanolate ≥97%
Aluminium tri-tert-butanolate
Very soluble in organic solvents
Purity: 97%
CAS No.: 556-91-2
Molecular Formula: Al[OC(CH3)3]3
Formula Weight: 246.32
Physical Form: Powder
Density: 1.0251
Melting Point: 200-202°
Boiling Point: 156°/2mm
Solubility: Very soluble in organic solvents
Beilstein Reference: 4148229
Merck Reference: 14,333
UN No.: 2925
MDL No.: MFCD00008801
Formula:
C₁₂H₂₇AlO₃ Boiling Pt: 156 °C (2 mmHg) Melting Pt: 200…202 °C Density: 1.025 Storage Temperature: Ambient |
MDL Number:
MFCD00008801 CAS Number: 556-91-2 EINECS: 209-146-1 UN: 2925 ADR: 4.1,III Merck Index: 14,333 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...