Calcium cyanamide, tech.
Supplier: Thermo Scientific Chemicals
|
Danger
|
89462-22
89462-36
CAAA89462-22EA
77.26
CAD
CAAA89462-22
CAAA89462-36
Calcium cyanamide, tech.
Calcium cyanamide
CAS No.: 156-62-7
Molecular Formula: CCaN2
Formula Weight: 80.11
Physical Form: Powder
Density: 2.29
Sensitivity: Moisture Sensitive
Appearance: Gray to dark gray
Odor: Specific type
Melting Point: 1340°
Beilstein Reference: 4124391
Merck Reference: 14,1662
UN No.: 1403
MDL No.: MFCD00064894
Formula:
CCaN₂ Melting Pt: 1340 °C Density: 2.29 Storage Temperature: Ambient |
MDL Number:
MFCD00064894 CAS Number: 156-62-7 EINECS: 205-861-8 UN: 1403 ADR: 4.3,III Merck Index: 14,1662 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...