Copper(II) iodate, anhydrous ≥95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Copper iodate

89459-04 89459-09 89459-18
CAAA89459-04EA 115.66 CAD
CAAA89459-04 CAAA89459-09 CAAA89459-18
Copper(II) iodate, anhydrous ≥95%
Copper (II) iodate

Purity: 95% CAS No.: 13454-89-2 Molecular Formula: Cu(IO3)2 Formula Weight: 413.35 Physical Form: Powder Density: 5.241 Melting Point: dec. UN No.: 1479 MDL No.: MFCD00016055

Formula: CuI₂O₆
MW: 413.35 g/mol
Melting Pt: Dec.
Density: 5.241
Storage Temperature: Ambient
MDL Number: MFCD00016055
CAS Number: 13454-89-2
UN: 1479
ADR: 5.1 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR