Sodium hexabromoplatinate(IV) hexahydrate

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Disodium hexabromoplatinate hexahydrate

88966-03 88966-06
CAAA88966-03EA 241.74 CAD
CAAA88966-03 CAAA88966-06
Sodium hexabromoplatinate(IV) hexahydrate
Disodium hexabromoplatinate

CAS No.: 39277-13-9 Molecular Formula: Na2PtBr6.6H2O Formula Weight: 828.49 (720.49anhy) Physical Form: Crystalline MDL No.: MFCD00014237

Formula: Na₂PtBr₆·6H₂O
MW: 828.49 g/mol (720.49 anhy)
Storage Temperature: Ambient
MDL Number: MFCD00014237
CAS Number: 39277-13-9
EINECS: 254-398-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR