Aluminium tributanolate 90+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
87754-09
87754-18
CAAA87754-09EA
237
CAD
CAAA87754-09
CAAA87754-18
Aluminium tributanolate 90+%
Aluminium tributanolate
Purity: 90+%
CAS No.: 3085-30-1
Molecular Formula: C12H27AlO3
Formula Weight: 246.32
Physical Form: Lump
Sensitivity: Moisture Sensitive
UN No.: 3181
MDL No.: MFCD00015237
Formula:
C₁₂H₂₇AlO₃ Melting Pt: 102 °C Storage Temperature: Ambient |
MDL Number:
MFCD00015237 CAS Number: 3085-30-1 EINECS: 221-394-2 UN: 3181 ADR: 4.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...