Copper(II) cyclohexanebutyrate Cu 15,8% for atomic absorption spectroscopy

Supplier: Thermo Scientific Chemicals

87262-03 87262-06
CAAA87262-03EA 48.51 CAD
CAAA87262-03 CAAA87262-06
Copper(II) cyclohexanebutyrate Cu 15,8% for atomic absorption spectroscopy
Copper (II) cyclohexanebutyrate

CAS No.: 2218-80-6 Molecular Formula: C20H34CuO4 Formula Weight: 402.01 Physical Form: Powder Melting Point: 126° dec. MDL No.: MFCD00036399

Formula: C₂₀H₃₄CuO₄
MW: 402.03
Melting Pt: 126 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00036399
CAS Number: 2218-80-6
EINECS: 218-723-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR