Potassium heptafluoroniobate ≥99.5%
Supplier: Thermo Scientific Chemicals
|
Danger
|
51110-09
51110-18
CAAA51110-09EA
63.04
CAD
CAAA51110-09
CAAA51110-18
Potassium heptafluoroniobate ≥99.5%
Potassium heptafluoroniobate
Purity: 99.5%
CAS No.: 16924-03-1
Molecular Formula: K2NbF7
Formula Weight: 304.10
Physical Form: Powder
Sensitivity: Hygroscopic
UN No.: 3288
MDL No.: MFCD00042368
Formula:
F₇K₂Nb Storage Temperature: Ambient |
MDL Number:
MFCD00042368 CAS Number: 16924-03-1 EINECS: 240-987-7 UN: 3288 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...