Boron (≥96% ¹⁰B) (metals basis), crystalline powder -60+100 mesh

Supplier: Thermo Scientific Chemicals
Warning

44256-06 44256-14
CAAA44256-06EA 86.9 CAD
CAAA44256-06 CAAA44256-14
Boron (≥96% ¹⁰B) (metals basis), crystalline powder -60+100 mesh
Boron

Neutron absorber


Purity: 99.9% CAS No.: 7440-42-8 Odor: Odorless UN No.: 3178 MDL No.: MFCD00143869

Formula: B
Density: 1.73 g/cm³ (15 °C)
Storage Temperature: Ambient
MDL Number: MFCD00134034
CAS Number: 7440-42-8
EINECS: 231-151-2
UN: 3178
ADR: 4.1,III
Merck Index: 13,01333

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR