Thulium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
44168-03
44168-06
CAAA44168-03EA
481.9
CAD
CAAA44168-03
CAAA44168-06
Thulium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Thulium
Purity: 99.9%
CAS No.: 7440-30-4
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Grey to silver
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011281
Formula:
Tm Boiling Pt: 1727 °C (1013 hPa) Melting Pt: 1544.85 °C Density: 9.318 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011281 CAS Number: 7440-30-4 EINECS: 231-140-2 UN: 3089 ADR: 4.1,II Merck Index: 12,09535 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...