Lutetium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Supplier: Thermo Scientific Chemicals
|
Danger
|
44167-03
44167-06
CAAA44167-03EA
613.04
CAD
CAAA44167-03
CAAA44167-06
Lutetium ≥99.9% (REO, rare earth oxide basis), powder -40 mesh
Lutetium
Purity: 99.9%
CAS No.: 7439-94-3
Physical Form: Packaged under argon
Sensitivity: Air and Moisture Sensitive
Appearance: Silver
Odor: Odorless
UN No.: 3089
MDL No.: MFCD00011098
Formula:
Lu Boiling Pt: 3000 °C (1013 hPa) Melting Pt: 1651…1675 °C Density: 9.849 g/cm³ (25 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011098 CAS Number: 7439-94-3 EINECS: 231-103-0 UN: 3089 ADR: 4.1,II Merck Index: 12,05642 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...